missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EB2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98106F
This item is not returnable.
View return policy
Description
EB2 Polyclonal antibody specifically detects EB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| EB2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| FITC | |
| APC-binding protein EB2, EB1, EB2APC-binding protein EB1, End-binding protein 2, microtubule-associated protein, RP/EB family, member 2, RP1microtubule-associated protein RP/EB family member 2, T-cell activation protein, EB1 family | |
| Produced in rabbits immunized with E. coli-derived Human EB2 fragment. | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| 10982 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction