missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EB2 (MAPRE2) Monoclonal Antibody (OTI1B10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502860S
This item is not returnable.
View return policy
Description
EB2 (MAPRE2) Monoclonal antibody specifically detects EB2 (MAPRE2) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| EB2 (MAPRE2) | |
| Monoclonal | |
| 0.37 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15555 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10982 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1B10 | |
| Unconjugated | |
| MAPRE2 | |
| AI314113; APC-binding protein EB1; APC-binding protein EB2; C820009F03Rik; CSCSC2; D18Abb1e; EB1; EB2; End-binding protein 2; Mapre2; microtubule associated protein RP/EB family member 2; microtubule-associated protein RP/EB family member 2; microtubule-associated protein, RP/EB family, member 2; RP1; T-cell activation protein, EB1 family; T-cell activation protein, EB1 family, member 2 | |
| Full length human recombinant protein of human MAPRE2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion