missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBAG9/RCAS1 Antibody (019), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90266H
This item is not returnable.
View return policy
Description
EBAG9/RCAS1 Monoclonal specifically detects EBAG9/RCAS1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| EBAG9/RCAS1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 19 | |
| Western Blot, ELISA | |
| Cancer-associated surface antigen RCAS1, EB9cancer associated surface antigen, estrogen receptor binding site associated, antigen, 9, Estrogen receptor-binding fragment-associated gene 9 protein, RCAS1PDAF, receptor-binding cancer antigen expressed on SiSo cells | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human EBAG9/RCAS1 (O00559; Arg28-Ser213). | |
| 0.1 mL | |
| Apoptosis | |
| 9166 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction