missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBAG9/RCAS1 Antibody (CPTC-EBAG9-2), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08860JF646
This item is not returnable.
View return policy
Description
EBAG9/RCAS1 Monoclonal antibody specifically detects EBAG9/RCAS1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| EBAG9/RCAS1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Cancer-associated surface antigen RCAS1, EB9cancer associated surface antigen, estrogen receptor binding site associated, antigen, 9, Estrogen receptor-binding fragment-associated gene 9 protein, RCAS1PDAF, receptor-binding cancer antigen expressed on SiSo cells | |
| Recombinant full-length human EBAG9/RCAS1 protein (Uniprot: O00559) | |
| 100 μg | |
| Apoptosis | |
| 9166 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| CPTC-EBAG9-2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction