missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBAG9/RCAS1 Antibody (rEBAG9/7264), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20399-100ug
This item is not returnable.
View return policy
Description
EBAG9/RCAS1 Monoclonal antibody specifically detects EBAG9/RCAS1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| EBAG9/RCAS1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Cancer-associated surface antigen RCAS1, EB9cancer associated surface antigen, estrogen receptor binding site associated, antigen, 9, Estrogen receptor-binding fragment-associated gene 9 protein, RCAS1PDAF, receptor-binding cancer antigen expressed on SiSo cells | |
| Recombinant full-length human EBAG9/RCAS1 protein | |
| 100 μg | |
| Apoptosis | |
| 9166 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rEBAG9/7264 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction