missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBAG9/RCAS1 Rabbit anti-Human, Clone: 19, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90266-50UL
This item is not returnable.
View return policy
Description
EBAG9/RCAS1 Monoclonal specifically detects EBAG9/RCAS1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| EBAG9/RCAS1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| 19 | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate | |
| Cancer-associated surface antigen RCAS1, EB9cancer associated surface antigen, estrogen receptor binding site associated, antigen, 9, Estrogen receptor-binding fragment-associated gene 9 protein, RCAS1PDAF, receptor-binding cancer antigen expressed on SiSo cells | |
| Recombinant Human EBAG9/RCAS1 Protein | |
| 50 μg | |
| Apoptosis | |
| 9166 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction