missing translation for 'onlineSavingsMsg'
Learn More

Ebastine, MedChemExpress

Product Code. 16452196
Encompass_Preferred
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 16452196

Brand: Medchem Express HYB0674

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ebastine (LAS-W 090) is an orally active, second-generation histamine H1 receptor antagonist. Ebastine can be used for the symptoms of allergic rhinitis and chronic idiopathic urticaria research.

Ebastine (LAS-W 090) is an orally active, second-generation histamine H1 receptor antagonist. Ebastine can be used for the symptoms of allergic rhinitis and chronic idiopathic urticaria research.

Specifications

Chemical Name or Material Ebastine
Physical Form Solid
CAS 90729-43-4
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C32H39NO2
For Use With (Application) COVID-19-immunoregulation
Synonym LAS-W 090 RP64305
Solubility Information Ethanol : 58.75 mg/mL (125.09 mM; Need ultrasonic) ∣DMSO : 8.33 mg/mL (17.74 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C(C)(C)C)C=C1)CCCN2CCC(OC(C3=CC=CC=C3)C4=CC=CC=C4)CC2
Molecular Weight (g/mol) 469.66
Formula Weight 469.66
Percent Purity 99.54%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.