missing translation for 'onlineSavingsMsg'
Learn More

EBF-1 Antibody (SR2090), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-22035-100ul

 View more versions of this product

Product Code. 18630734

  • € 449.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

EBF-1 Monoclonal antibody specifically detects EBF-1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EBF-1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
1879
Store at -20° C. Avoid freeze/thaw cycles.
IgG
Western Blot
SR2090
Western Blot 1:500-1:1000
COE1, Collier, Olf and EBF transcription factor 1, Early B-cell factor, early B-cell factor 1, EBFFLJ41763, O/E-1, OE-1, OLF1FLJ39389, olfactory neuronal transcription factor 1, transcription factor COE1
A synthesized peptide derived from human EBF-1 (Uniprot #: Q9UH73)
100 μg
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.