missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EBF1 Polyclonal Antibody

Product Code. 17159433
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17159433 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17159433 Supplier Invitrogen™ Supplier No. PA5111517

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MFGIQESIQR SGSSMKEEPL GSGMN Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

Transcriptional activator which recognizes variations of the palindromic sequence 5'-ATTCCCNNGGGAATT-3'. Target protein is a transcription factor that may regulate several olfactory-specific genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBF1
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EBF1
Gene Accession No. Q9UH73
Gene Alias COE1; Collier; Collier, Olf and EBF transcription factor 1; early B cell factor 1; early B-cell factor; early B-cell factor (olfactory neuronal transcription factor 1); early B-cell factor 1; Ebf; EBF transcription factor 1; EBF1; O/E-1; OE-1; OLF1; Olf-1; Olfactory neuronal transcription factor; olfactory neuronal transcription factor 1; transcription factor COE1
Gene Symbols EBF1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human EBF1. Recombinant protein control fragment (Product #RP-105171).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1879
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.