missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EBF2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15548382
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15548382 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548382 Supplier Invitrogen™ Supplier No. PA547890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) EBF-1 and rmEBF-3 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

Early B-cell factor 2 (EBF2) is a gene located on chromosome 8p11.23, encoding a transcription factor involved in the regulation of gene expression during neural and adipose tissue development. EBF2 belongs to the COE family of transcription factors and plays a pivotal role in cellular differentiation processes. In the nervous system, EBF2 is essential for the development and function of neurons, particularly in regions like the olfactory system. Additionally, EBF2 is involved in adipogenesis, where it regulates genes responsible for the differentiation of brown adipocytes, contributing to thermogenesis and energy metabolism. Dysfunction or altered expression of EBF2 can impact neurological function and metabolic homeostasis, potentially influencing conditions such as obesity, diabetes, and neurodevelopmental disorders. Research into EBF2 is exploring its role in these physiological contexts and the implications of its regulatory mechanisms, offering insights into potential therapeutic approaches.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBF2
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene EBF2
Gene Accession No. O08792, Q9HAK2
Gene Alias COE2; Collier, Olf and EBF 2; D14Ggc1e; early B cell factor 2; early B-cell factor 2; Ebf2; EBF-2; metencephalon-mesencephalnon-olfactory transcription factor 1; metencephalon-mesencephalon-olfactory transcription factor 1; MET-mesencephalon-olfactory TF1; MET-mesencephalon-olfactory transcription factor 1; Mmot1; O/E-3; OE-3; Olf-1/EBF-like 3; transcription factor COE2
Gene Symbols EBF2
Host Species Sheep
Immunogen E. coli-derived recombinant mouse EBF-2 Arg407-Ser519.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13592, 64641
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.