missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI2/GPR183 Antibody (486633) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10272JF669
This item is not returnable.
View return policy
Description
EBI2/GPR183 Monoclonal antibody specifically detects EBI2/GPR183 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| EBI2/GPR183 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EBI2Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupledreceptor), EBV-induced G-protein coupled receptor 2, Epstein-Barr virus-induced G-protein coupled receptor 2, G protein-coupled receptor 183, G-protein coupled receptor 183 | |
| Mouse myeloma cell line NS0-derived transfected withrecombinant human EBI2, Accession # P32249 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 486633 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR, Immune System Diseases, Immunology | |
| 1880 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction