missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Antibody (10J803) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-03940JF525
This item is not returnable.
View return policy
Description
EBI3 Monoclonal antibody specifically detects EBI3 in Mouse samples. It is validated for Flow Cytometry,Western Blot
Specifications
| EBI3 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
| Full-length recombinant mouse EBI3 protein | |
| 0.1 mL | |
| Apoptosis, Cancer, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| 10148 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, Western Blot | |
| 10J803 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction