missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Antibody (15k8D10) - Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
€ 493.00 - € 2090.00
Specifications
| Antigen | IL-27/IL-35 EBI3 Subunit |
|---|---|
| Clone | 15k8D10 |
| Dilution | Western Blot 3-5ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18473862
|
Novus Biologicals
NBP2-27363-1mg |
1 mg |
€ 2090.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776723
|
Novus Biologicals
NBP2-27363 |
€ 493.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18157043
|
Novus Biologicals
NBP2-27363-0.5MG |
0.5 mg |
€ 1305.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EBI3 Monoclonal specifically detects EBI3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IL-27/IL-35 EBI3 Subunit | |
| Western Blot 3-5ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 10148 | |
| A full-length human Interleukin-35-Fc fusion protein was used as immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 15k8D10 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
| EBI3 | |
| IgG1 κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title