missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Antibody (15k8D10), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27363IR
This item is not returnable.
View return policy
Description
EBI3 Monoclonal specifically detects EBI3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-27/IL-35 EBI3 Subunit | |
| Monoclonal | |
| DyLight 755 | |
| NP_000873.2 | |
| EBI3 | |
| A full-length human Interleukin-35-Fc fusion protein was used as immunogen for this antibody. | |
| 0.1 mL | |
| Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| 10148 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 15k8D10 | |
| Western Blot, Immunohistochemistry-Paraffin | |
| cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The human Interleukin-35 protein is a dimeric protein consisting of the human IL-12a/p35 subunit (NP_000873.2) and the human EBI3 (NP_005746) protein. Clone 15K8D10 tests as being specific to the EBI3 subunit of the target protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction