missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Antibody (42N1G4) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-27356CL1
This item is not returnable.
View return policy
Description
EBI3 Monoclonal antibody specifically detects EBI3 in Human samples. It is validated for Flow Cytometry,Western Blot
Specifications
| EBI3 | |
| Monoclonal | |
| CoraFluor 1 | |
| cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
| Full-length recombinant human EBI3 protein full length recombinant human EBI3 (cat # 100-162) | |
| 0.1 mL | |
| Apoptosis, Cancer, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| 10148 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Flow Cytometry, Western Blot | |
| 42N1G4 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction