missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Antibody (42N1G4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27356F
This item is not returnable.
View return policy
Description
EBI3 Monoclonal specifically detects EBI3 in Human samples. It is validated for Flow Cytometry.
Specifications
| IL-27/IL-35 EBI3 Subunit | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| EBI3 | |
| Full-length recombinant human EBI3 protein full length recombinant human EBI3 (cat # 100-162) | |
| 0.1 mL | |
| Apoptosis, Cancer, Chemokine Research, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| 10148 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Flow Cytometry | |
| 42N1G4 | |
| Flow Cytometry | |
| cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction