missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBI3 Antibody (607201), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6456CL1
This item is not returnable.
View return policy
Description
EBI3 Monoclonal antibody specifically detects EBI3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| EBI3 | |
| Monoclonal | |
| CoraFluor 1 | |
| cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta | |
| Mouse myeloma cell line NS0-derived recombinant human IL-27/IL-35 EBI3 Subunit, Arg21-Lys229 (predicted), Accession # Q14213 | |
| 0.1 mL | |
| Apoptosis, Cancer, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction | |
| 10148 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 607201 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction