Learn More
EBLN2 Monoclonal Antibody (OTI1A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808873S
Encompass_Preferred
 View more versions of this product
Product Code. 17580594
  • € 242.00 / 30µL
Description
EBLN2 Monoclonal antibody specifically detects EBLN2 in Human samples. It is validated for Western Blot
Specifications
Specifications
EBLN2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q6P2I7
Mouse
Affinity chromatography
RUO
55096
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1A2
Unconjugated
EBLN2
EBLN-1; EBLN2; EBLN-2; endogenous Borna-like N element-1; endogenous Borna-like N element-2; Endogenous Bornavirus-like nucleoprotein 2; GK006; ikappaBalpha; I-kappa-B-alpha; Ikba; ikB-alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells, alpha; NF-kappa-B inhibitor alpha; Nfkbia; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, alpha; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha; RL/IF-1
Full length human recombinant protein of human EBLN2 produced in E.coli.