missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBLN2 Monoclonal Antibody (OTI2D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808871S
This item is not returnable.
View return policy
Description
EBLN2 Monoclonal antibody specifically detects EBLN2 in Human samples. It is validated for Western BlotSpecifications
| EBLN2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NYD6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3226 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2D5 | |
| Unconjugated | |
| HOXC10 | |
| EBLN-1; EBLN2; EBLN-2; endogenous Borna-like N element-1; endogenous Borna-like N element-2; Endogenous Bornavirus-like nucleoprotein 2; GK006; ikappaBalpha; I-kappa-B-alpha; Ikba; ikB-alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells, alpha; NF-kappa-B inhibitor alpha; Nfkbia; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, alpha; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha; RL/IF-1 | |
| Human recombinant protein fragment corresponding to amino acids 1-262 of human HOXC10 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction