missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBLN2 Monoclonal Antibody (OTI2D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808871
This item is not returnable.
View return policy
Description
May act as an RNA-binding protein. The C-terminal region is highly homologous to the bornavirus nucleocapsid N protein that binds viral RNA and oligomerizes. The viral protein also possesses a nuclear import and a nuclear export signal. These 2 signals seem absent in EBLN-2 supporting an unrelated function in Human.Specifications
| EBLN2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6P2I7 | |
| EBLN2 | |
| Full length human recombit protein of human EBLN2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2D5 | |
| Unconjugated | |
| EBLN2 | |
| EBLN-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55096 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction