Learn More
EBLN2 Mouse anti-Human, Clone: OTI3E1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF808875
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May act as an RNA-binding protein. The C-terminal region is highly homologous to the bornavirus nucleocapsid N protein that binds viral RNA and oligomerizes. The viral protein also possesses a nuclear import and a nuclear export signal. These 2 signals seem absent in EBLN-2 supporting an unrelated function in Human.Specifications
| EBLN2 | |
| Monoclonal | |
| Unconjugated | |
| EBLN2 | |
| EBLN-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55096 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI3E1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q6P2I7 | |
| EBLN2 | |
| Full length human recombit protein of human EBLN2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.