Learn More
EBNA1 Antibody (E1-2.5), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB500-312FR
Product Code. 18177269
  • € 548.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
EBNA1 Monoclonal specifically detects EBNA1 in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
EBNA1
Monoclonal
DyLight 680
BKRF1, Epstein-Barr nuclear antigen 1, HHV4gp098, nuclear antigen EBNA1
Synthetic peptide corresponding to the repetitive Gly-Ala region of EBNA-1.
0.1 mL
Primary
Virus
Purified
ELISA, Immunocytochemistry, Immunofluorescence
E1-2.5
ELISA, Immunocytochemistry/Immunofluorescence
Mouse
Protein A purified
RUO
The antibody E1-2.5 recognizes repetitive Gly-Ala region of the Epstein-Barr virus (EBV) nuclear antigen 1 (EBNA-1). EBNA-1 is the only viral protein consistently expressed in all malignancies associated with EBV.
Store at 4C in the dark.
IgG2b