missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBOV GP Antibody (06), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06458C
This item is not returnable.
View return policy
Description
EBOV GP Monoclonal antibody specifically detects EBOV GP in Virus samples. It is validated for ELISA
Specifications
| EBOV GP | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3160774 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 06 | |
| ELISA | |
| second secreted glycoprotein, small secreted glycoprotein, Spike glycoprotein | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ebola virus EBOV (subtype Zaire, strain Mayinga 1976) Glycoprotein/ GP (AAC54887.1; Met1-Gln650). | |
| 0.1 mL | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction