missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
EBOV GP Antibody (06), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06458F
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
EBOV GP Monoclonal antibody specifically detects EBOV GP in Virus samples. It is validated for ELISA
Tekniske data
| EBOV GP | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3160774 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 06 | |
| ELISA | |
| second secreted glycoprotein, small secreted glycoprotein, Spike glycoprotein | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Ebola virus EBOV (subtype Zaire, strain Mayinga 1976) Glycoprotein/ GP (AAC54887.1; Met1-Gln650). | |
| 0.1 mL | |
| Primary | |
| Virus | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion