missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBOV Nucleoprotein Antibody (09), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-12759AF405
This item is not returnable.
View return policy
Description
EBOV Nucleoprotein Monoclonal antibody specifically detects EBOV Nucleoprotein in Ebola virus samples. It is validated for Western Blot
Specifications
| EBOV Nucleoprotein | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Ebola virus | |
| Purified |
| Western Blot | |
| 09 | |
| 50mM Sodium Borate | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant EBOV Nucleoprotein (Accession#: AHX24646.1; His630-Gln739). | |
| 100 μg | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction