Learn More
EBPL Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-05841PE
Product Code. 18689958
  • € 407.00 / 0.10mL
Description
EBPL Polyclonal antibody specifically detects EBPL in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Rabbit
84650
PBS
Produced in rabbits immunized with E. coli-derived Human EBPL fragment.
Store at 4C in the dark.