Learn More
EBV Latent Membrane Protein 1 Antibody (LMPO24) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50383
 View more versions of this product
Product Code. 18606636
  • € 381.00 / 0.10mL
Description
EBV Latent Membrane Protein 1 Monoclonal antibody specifically detects EBV Latent Membrane Protein 1 in Virus samples. It is validated for Flow Cytometry, Immunohistochemistry, Flow (Intracellular), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EBV Latent Membrane Protein 1
Monoclonal
1 mg/mL
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Flow (Intracellular) 1:10 - 1:1000, CyTOF-ready
Mouse
Protein A purified
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Flow Cytometry, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
LMPO24
Unconjugated
PBS
Purified plasma membranes from EBV Latent Membrane Protein 1 expressing insect cells infected with a recombinant EBV Latent Membrane Protein 1 baculovirus
0.1 mL
Cancer, Cellular Signaling, Immunology, Virology Bacteria and Parasites
Virus