Learn More
Elma™ Clean 70 Concentrated Cleaner
Product Code. 10059420
Change view
Click to view available options
Volume (Metric):
1 L
25 L
Unit Size:
1 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10059420 1 L 1 L
11706279 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10059420 Supplier Elma™ Supplier No. 5810300000
Excellent for the thorough cleaning of laboratory equipment, tools and instruments made of glass, ceramics, alkaline-stable plastic, rubber, metal and light metal. Elma™ Clean 70 Concentrated Cleaner is an excellent choice for universal lab cleaning.
  • Available in a range of volumes
  • Removes dust, fingerprints, pigments, grease and oil
Specifications
Product Type EC 70 Cleaner
Form Liquid
pH 13.3
Appearance Beige
Boiling Point >100°C
Certifications/Compliance Annex VII A, EC 648/2004, UN 1719
Composition Anionic ten sides 5 to 15%, amphoteric ten sides <5%, Phosphates 5 to 15%, NTA <5%
Concentration Ultrasonic bath 1 to 5%, without ultrasound 5 to 10%
Container Type Bottle
Content And Storage Above -5°C
For Use With (Application) For clean laboratory instruments, equipment and items made from Glass, Stainless Steel, Ceramics, Alkaline resistant Plastics, Rubber and light Metals
Volume (Metric) 1 L
Show More Show Less
compliance-icons
Product Identifier
  • elma clean 70 (EC 70)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.