missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ECD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581542
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse ECD.
Regulator of p53/TP53 stability and function. Inhibits MDM2-mediated degradation of p53/TP53 possibly by cooperating in part with TXNIP (PubMed:16849563, PubMed:23880345). May be involved transcriptional regulation. In vitro has intrinsic transactivation activity enhanced by EP300. May be a transcriptional activator required for the expression of glycolytic genes (PubMed:19919181, PubMed:9928932). Involved in regulation of cell cycle progression. Proposed to disrupt Rb-E2F binding leading to transcriptional activation of E2F proteins (PubMed:19640839). The cell cycle -regulating function may depend on its RUVBL1-mediated association with the R2TP complex (PubMed:26711270). May play a role in regulation of pre-mRNA splicing (PubMed:24722212). [UniProt].
Specifications
| ECD | |
| Polyclonal | |
| Unconjugated | |
| ECD | |
| 5730461K03Rik; Ecd; ecdysoneless cell cycle regulator; ecdysoneless homolog; ecdysoneless homolog (Drosophila); GCR2; HSGT1; Human suppressor of GCR two; protein ecdysoneless homolog; protein SGT1; protein SGT1 homolog; SGT1; SGT1 protein; suppressor of GCR2; suppressor of S. cerevisiae gcr2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 70601 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9CS74 | |
| ECD | |
| A synthetic peptide corresponding to the center region of the Mouse ECD/Ecdysoneless homolog. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction