missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECE-1 Antibody (303913), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB17841L
This item is not returnable.
View return policy
Description
ECE-1 Monoclonal antibody specifically detects ECE-1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| ECE-1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1889 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 303913 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| EC 3.4.24, EC 3.4.24.71, ECE, ECE-1, endothelin converting enzyme 1, endothelin-converting enzyme 1 | |
| Mouse myeloma cell line NS0-derived recombinant human ECE-1, Gln90-Trp770, Accession # P42892 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction