Learn More
Invitrogen™ ECE2 Monoclonal Antibody (233914)
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA523941
Product Code. 15543632
  • € 475.00 / 100µg
Description
In Western blots, 30% cross-reactivity with recombinant human (rh) ECE-1 and no cross-reactivity with rhKELL is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ECE2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P0DPD6
Ece2
Mouse myeloma cell line NS0-derived recombinant human ECE-2 Val104-Leu787.
100 μg
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
233914
Unconjugated
Ece2
1810009K13Rik; 6330509A19Rik; 9630025D12Rik; BB127715; Ece2; ECE-2; ECE-2a-1; ECE-2a-2; ECE-2b-1; ECE-2b-2; EEF1A lysine methyltransferase 4; EEF1AKMT4; EEF1AKMT4-ECE2; EEF1AKMT4-ECE2 readthrough transcript protein; endothelin converting enzyme 2; endothelin converting enzyme-2; endothelin-converting enzyme 2; Endothelin-converting enzyme 2 region; endothelin-converting enzyme 2a-1; endothelin-converting enzyme 2a-2; endothelin-converting enzyme 2b-1; endothelin-converting enzyme 2b-2; KIAA0604; Methyltransferase-like region; UNQ403/PRO740
Rat
Protein A/G
RUO
9718
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized