Learn More
ECH1 Antibody (002), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90288APC
This item is not returnable.
View return policy
Description
ECH1 Monoclonal antibody specifically detects ECH1 in Human samples. It is validated for Western Blot, ELISA