missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECHS1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-96977UV
This item is not returnable.
View return policy
Description
ECHS1 Polyclonal specifically detects ECHS1 in Human samples. It is validated for Western Blot.
Specifications
| ECHS1 | |
| Polyclonal | |
| Western Blot | |
| EC 4.2.1, enoyl CoA hydratase, short chain, 1, mitochondrial, enoyl Coenzyme A hydratase, short chain, 1, mitochondrial, Enoyl-CoA hydratase 1, enoyl-CoA hydratase, mitochondrial, SCEHEC 4.2.1.17, Short-chain enoyl-CoA hydratase | |
| Produced in rabbits immunized with purified, recombinant ECHS1 (AAH08906.1; 28A-290Q). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 1892 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction