Learn More
BD Eclipse™ Hypodermic Injection Needle with SmartSlip™ Technology
Product Code. 12577906
Click to view available options
Needle Gauge:
21 G
Unit Size:
Pack of 100
Product Code. 12577906
Brand: BD 305895
Ensures protection from needlestick injuries after injection. The BD Eclipse™ Hypodermic Injection Needle features the same safety mechanism as the proven BD Eclipse™ Blood Collection Needle, with SmartSlip™ Technology to ensure a secure connection to Luer slip syringes.
  • Unique SmartSlip technology helps to ensure a secure connection to Luer slip syringes
  • Bevel oriented to safety cover accommodates low angle injections
  • One technology for skin injection and blood collection
  • Requires minimal change in injection technique
  • The hub incorporates a plastic clip to encourage user to attach needle more securely to Luer slip syringe
  • Wide textured finger pad provides safe activation area for thumb or forefinger
  • Integral safety cover and extended sidewall to enhance protection
  • Needle remains locked inside activated needle cover
  • Activates with a touch of a finger – no hard surface required for activation
  • Colour coded hub for easy needle size identification
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU directive
Color Green
Needle Gauge 21 G
Diameter (Metric) Outer Needle 0.8 mm
For Use With (Application) Hypodermic injections
Includes Safety cover
Length (Metric) Needle 40 mm
Needle Point Style Bevel
For Use With (Equipment) Luer Slip Syringe
Connection Type Luer