Learn More
Bellingham Stanley™ Eclipse Professional Grade Optical Hand-Held AntiFreeze Refractometer
missing translation for 'orderingAttributeHoverText'
Förpackningsstorlek
Each
Tekniske data
Tekniske data
| Type | Eclipse Professional Grade Optical Hand Held Refractometer |
| Scale Range | 0°C to -40°C Antifreeze Protection, 1.1 to 1.35 Battery Acid Specific Gravity |
| For Use With (Application) | ‘On the spot’ determination of Refractive Index (RI) of liquids or the concentration of solutions |
| IP Rating | IP65 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion