missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECM-1/Secretory Component P85 Antibody (ECM1/2889R), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08458R
This item is not returnable.
View return policy
Description
ECM-1/Secretory Component P85 Monoclonal antibody specifically detects ECM-1/Secretory Component P85 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ECM-1/Secretory Component P85 | |
| Monoclonal | |
| DyLight 550 | |
| ECM1, extracellular matrix protein 1, Secretory Component Glycoprotein, Secretory component p85 | |
| Recombinant human full-length ECM-1/Secretory Component P85 protein (Uniprot: Q16610) | |
| 100 μg | |
| Extracellular Matrix, Immunology | |
| 1893 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| ECM1/2889R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction