Learn More
Lauda™ Eco Boiling Water Bath
Specifications
Specifications
| Capacity (Metric) | 6 L |
| Temperature Range | 100°C |
| Type | Boiling Water Bath |
| For Use With (Application) | Used to heat small volumes of water in laboratories |
| Material | Polycarbonate |