missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECRG4 Antibody (OTI3B10), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72294UV
This item is not returnable.
View return policy
Description
ECRG4 Monoclonal specifically detects ECRG4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| ECRG4 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| C2orf40 | |
| Full length recombinant protein of human ECRG4 (NP_115787) produced in HEK293T cells. | |
| 0.1 mL | |
| 84417.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI3B10 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| chromosome 2 open reading frame 40, ECRG4augurin, esophageal cancer related gene 4 protein, Esophageal cancer-related gene 4 protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction