Learn More
Invitrogen™ Ofatumumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559578
 View more versions of this product
Product Code. 30282565
  • € 1204.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ofatumumab is a fully human monoclonal antibody to CD20, which appears to inhibit early-stage B lymphocyte activation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ofatumumab
Recombinant Monoclonal
Unconjugated
HuMax-CD20
Protein A
RUO
Human
Antibody
IgG1 κ
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 0.01% Tween 80, 8% sucrose and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized