missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EDA-DA
GREENER_CHOICE

Product Code. 18796156
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18796156 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18796156 Supplier Tocris Bioscience™ Supplier No. 7714/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Unnatural dipeptide building block with alkyne group for functionalizing peptidoglycan

EDA-DA is an unnatural dipeptide building block (an ethynyl-D-alanine and a D-alanine). It incorporates a biorthogonal alkyne group into peptidoglycan (PG) through MurF in the cytoplasmic pathway, which enables selective labeling via a click-chemistry reaction. EDA-DA allows labeling of PG in Gram-positive (B. subtilis), Gram-negative (E. coli and C. trachomatis), Mycobacterium (M. smegmatis) and moss plastids (P. patens) with azide modified fluorescent dyes such as Alexa Fluor 488.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(4,4,5,5-Tetradehydro-D-norvalyl)-D-alanine trifluoroacetate
CAS 87156-01-2
Quantity 10 mg
Target Unnatural Amino Acids
Molecular Formula C8H12N2O3.CF3CO2H
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.