Learn More
EDA2R/TNFRSF27/XEDAR Antibody (072), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90000CL1
This item is not returnable.
View return policy
Description
EDA2R/TNFRSF27/XEDAR Monoclonal antibody specifically detects EDA2R/TNFRSF27/XEDAR in Human samples. It is validated for ELISA
Specifications
| EDA2R/TNFRSF27/XEDAR | |
| Monoclonal | |
| CoraFluor 1 | |
| ectodysplasin A2 receptor, EDA-A2 receptor, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human EDA2R/TNFRSF27/XEDAR (Accession#: NP_068555.1; Met1-Thr138). | |
| 0.1 mL | |
| Signal Transduction | |
| 60401 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 072 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |