missing translation for 'onlineSavingsMsg'
Learn More

EDAR Antibody (19E8), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-42695AF405

Product Code. 18182819

  • € 527.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

EDAR Monoclonal specifically detects EDAR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EDAR
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
EDAR
Recombinant human EDAR (27-448aa) purified from E.coli.
100 μL
Apoptosis
10913
Store at 4C in the dark.
IgG2a κ
Western Blot, Flow Cytometry, ELISA
1900000000
Western Blot, Flow Cytometry, ELISA
Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.