missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDAR Antibody (19E8), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42695F
This item is not returnable.
View return policy
Description
EDAR Monoclonal specifically detects EDAR in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications
| EDAR | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| EDAR | |
| Recombinant human EDAR (27-448aa) purified from E.coli. | |
| 100 μL | |
| Apoptosis | |
| 10913 | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Flow Cytometry, ELISA | |
| 1900000000 | |
| Flow Cytometry, ELISA | |
| Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction