missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDAR Antibody (EDAR12), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-81103CL1
This item is not returnable.
View return policy
Description
EDAR Monoclonal antibody specifically detects EDAR in Canine,Chicken,Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Surface Plasmon Resonance,Neutralization,Immunocytochemistry/Immunofluorescence
Specifications
| EDAR | |
| Monoclonal | |
| CoraFluor 1 | |
| Anhidrotic ectodysplasin receptor 1, DLED3, Ectodermal dysplasia receptor, ectodysplasin 1, anhidrotic receptor, ectodysplasin A receptor, Ectodysplasin-A receptor, ED1R, ED5, EDA1R, EDA3EDA-A1 receptor, EDA-A1R, Edar, FLJ94390, HRM1, mouse, homolog of, tumor necrosis factor receptor superfamily member EDAR | |
| EDAR12 was prepared by immunixzing female OVE1B mice (with the Edar gene deleted) subcutaneously with mouse EDAR-Fc and positive hybridoma clones were screened for binding mEDAR by ELISA. | |
| 0.1 mL | |
| Apoptosis | |
| 10913 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| ELISA, Western Blot, Surface Plasmon Resonance, Neutralization, Immunocytochemistry/Immunofluorescence | |
| EDAR12 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Canine, Chicken, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction