missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDARADD Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92286-0.02ml
This item is not returnable.
View return policy
Description
EDARADD Polyclonal antibody specifically detects EDARADD in Mouse, Rat samples. It is validated for Western Blot
Specifications
| EDARADD | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| crinkled homolog, ectodysplasia A receptor associated death domain, ectodysplasin A receptor associated adapter protein, ectodysplasin-A receptor-associated adapter protein, ED3, EDA3, EDAR-associated death domain, EDAR-associated death domain protein, Protein crinkled homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-215 of human EDARADD (NP_665860.2). MGLRTTKQMGRGTKAPGHQEDHMVKEPVEDTDPSTLSFNMSDKYPIQDTELPKAEECDTITLNCPRNSDMKNQGEENGFPDSTGDPLPEISKDNSCKENCTCSSCLLRAPTISDLLNDQDLLDVIRIKLDPCHPTVKNWRNFASKWGMSYDELCFLEQRPQSPTLEFLLRNSQRTVGQLMELCRLYHRADVEKVLRRWVDEEWPKRERGDPSRHF | |
| 0.02 mL | |
| Cell Biology, Signal Transduction | |
| 128178 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction