missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EDEM1 Polyclonal Antibody

Product Code. 16386994
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16386994 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16386994 Supplier Invitrogen™ Supplier No. PA578296

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Jurkat, Jurkat membrane extract Predicted Reactivity: Mouse (87%), Rat (87%), Rhesus Monkey (100%), Bovine (92%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

EDEM1 belongs to the glycosyl hydrolase 47 family. It extracts misfolded glycoproteins, but not glycoproteins undergoing productive folding, from the calnexin cycle. It is directly involved in endoplasmic reticulum-associated degradation (ERAD) and targets misfolded glycoproteins for degradation in an N-glycan-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EDEM1
Applications Western Blot
Classification Polyclonal
Concentration 1.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene EDEM1
Gene Accession No. Q92611
Gene Alias A130059K23Rik; EDEM; EDEM1; ER degradation enhancer, mannosidase alpha-like 1; ER degradation enhancing alpha-mannosidase like protein 1; ER degradation ER degradation enhancer, mannosidase alpha-like 1; ER degradation-enhancing alpha-mannosidase-like 1; ER degradation-enhancing alpha-mannosidase-like protein 1; KIAA0212; mKIAA0212; RGD1563633
Gene Symbols EDEM1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human EDEM1. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9695
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.