missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDEM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568189
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
Involved in endoplasmic reticulum-associated degradation (ERAD). Accelerates the glycoprotein ERAD by proteasomes, by catalyzing mannose trimming from Man8GlcNAc2 to Man7GlcNAc2 in the N-glycans. Seems to have alpha 1,2-mannosidase activity.
Specifications
| EDEM3 | |
| Polyclonal | |
| Unconjugated | |
| EDEM3 | |
| 2310050N11Rik; Alpha-1,2-mannosidase EDEM3; C1orf22; EDEM3; ER degradation enhancer, mannosidase alpha-like 3; ER degradation enhancing alpha-mannosidase like protein 3; ER degradation-enhancing alpha-mannosidase-like 3; ER degradation-enhancing alpha-mannosidase-like protein 3; ER degradation-enhancing -mannosidase-like protein 3; RGD1561496 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 66967, 80267 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q2HXL6, Q9BZQ6 | |
| EDEM3 | |
| Synthesized peptide derived from human EDEM3. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction