missing translation for 'onlineSavingsMsg'
Learn More

EDF1 Monoclonal Antibody (OTI7B6), TrueMAB™, OriGene

Product Code. p-200015336
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17554214 30 μL 30µL
17534214 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17554214 Supplier Origene Technologies Supplier No. TA810774S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

EDF1 Monoclonal antibody specifically detects EDF1 in Human samples. It is validated for Western Blot

Specifications

Antigen EDF1
Applications Western Blot
Classification Monoclonal
Clone OTI7B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Aqp8
Gene Accession No. O94778
Gene Alias 0610008L11Rik; AA409425; calmodulin-associated peptide 19; calmodulin-associated peptide-19; CAP-19; CFAP280; Edf1; EDF-1; endothelial differentiation related factor 1; endothelial differentiation-related factor 1; Endothelial differentiation-related factor 1 homolog; hypothetical protein 1-9; hypothetical protein MGC73192; MBF1; MGC9058; multiprotein bridging factor 1; multiprotein-bridging factor 1; zgc:73192; zgc:86829
Host Species Mouse
Immunogen Full length human recombinant protein of human AQP8 produced in HEK293T.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.