Learn More
EDF1 Monoclonal Antibody (OTI7D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810789S
Encompass_Preferred
 View more versions of this product
Product Code. 17574254
  • € 242.00 / 30µL
Description
EDF1 Monoclonal antibody specifically detects EDF1 in Human samples. It is validated for Western Blot
Specifications
Specifications
EDF1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P09919
Mouse
Affinity chromatography
RUO
1440
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI7D9
Unconjugated
Csf3
0610008L11Rik; AA409425; calmodulin-associated peptide 19; calmodulin-associated peptide-19; CAP-19; CFAP280; Edf1; EDF-1; endothelial differentiation related factor 1; endothelial differentiation-related factor 1; Endothelial differentiation-related factor 1 homolog; hypothetical protein 1-9; hypothetical protein MGC73192; MBF1; MGC9058; multiprotein bridging factor 1; multiprotein-bridging factor 1; zgc:73192; zgc:86829
Full length human recombinant protein of human CSF3 produced in HEK293T.