missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EDF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583280
This item is not returnable.
View return policy
Description
Immunogen sequence: KLDRETEELH HDRVTLEVGK VIQQGRQSKG LTQKDLATKI NEKPQVIADY ESGRAIPNNQ VLGKIERAIG LKLRGKDIGK PIE Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.
This gene is known to suppress G-protein and mitogen-activated signal transduction in mammalian cells. The encoded protein shares significant similarity with Arabidopsis FUS6, which is a regulator of light-mediated signal transduction in plant cells. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| EDF1 | |
| Polyclonal | |
| Unconjugated | |
| EDF1 | |
| 0610008L11Rik; AA409425; calmodulin-associated peptide 19; calmodulin-associated peptide-19; CAP-19; CFAP280; Edf1; EDF-1; endothelial differentiation related factor 1; endothelial differentiation-related factor 1; Endothelial differentiation-related factor 1 homolog; hypothetical protein 1-9; hypothetical protein MGC73192; MBF1; MGC9058; multiprotein bridging factor 1; multiprotein-bridging factor 1; zgc:73192; zgc:86829 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8721 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60869 | |
| EDF1 | |
| Recombinant protein corresponding to Human EDF1. Recombinant protein control fragment (Product #RP-96551). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction